Assay Details
Gene Symbol: Dhodh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008553.1 | NP_001008553.1 | 1 - 2 | 75 | 80 | |
XM_006255601.2 | XP_006255663.1 | 1 - 2 | 248 | 80 | ||
XM_008772529.1 | XP_008770751.1 | 1 - 2 | 248 | 80 | ||
GenBank mRNA | X80778.1 | - | 1 - 2 | 75 | 80 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001008553.1, X80778.1, XM_006255601.2, XM_008772529.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
