Assay Details
Gene Symbol: Dhodh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008553.1 | NP_001008553.1 | 8 - 9 | 1181 | 134 | |
XM_006255601.2 | XP_006255663.1 | 8 - 9 | 1354 | 134 | ||
XM_008772529.1 | XP_008770751.1 | 8 - 9 | 1354 | 134 | ||
GenBank mRNA | X80778.1 | - | 8 - 9 | 1181 | 134 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
