Assay Details
Gene Symbol: Rad23a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013190.1 | NP_001013208.1 | 3 - 4 | 473 | 65 | |
XM_006255276.2 | XP_006255338.1 | 3 - 4 | 534 | 65 | ||
XM_006255277.2 | XP_006255339.1 | 3 - 4 | 534 | 65 | ||
XM_008772411.1 | XP_008770633.1 | 3 - 4 | 1789 | 65 | ||
GenBank mRNA | BC084695.1 | - | 3 - 4 | 473 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
