Assay Details
Gene Symbol: Cbfa2t3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108453.1 | NP_001101923.1 | 3 - 4 | 397 | 103 | |
XM_006255772.2 | XP_006255834.1 | 3 - 4 | 552 | 103 | ||
XM_008772629.1 | XP_008770851.1 | 3 - 4 | 2067 | 103 | ||
XM_008772631.1 | XP_008770853.1 | 3 - 4 | 552 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
