Assay Details
Gene Symbol: Prkaa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023991.1 | NP_076481.1 | 4 - 5 | 480 | 107 | |
XM_008763901.1 | XP_008762123.1 | 3 - 4 | 2209 | 107 | ||
GenBank mRNA | U12149.1 | - | 4 - 5 | 665 | 107 | |
Z29486.1 | - | 4 - 5 | 480 | 107 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
