Assay Details
Gene Symbol: Alg6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033709.1 | NP_001028881.1 | 6 - 7 | 621 | 90 | |
GenBank mRNA | BC101850.1 | - | 6 - 7 | 621 | 90 | |
FQ210318.1 | - | 6 - 7 | 636 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
