Assay Details
Gene Symbol: Elavl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006238385.2 | XP_006238447.1 | 6 - 7 | 814 | 87 | |
XM_006238388.2 | XP_006238450.1 | 5 - 6 | 617 | 87 | ||
XM_006238389.2 | XP_006238451.1 | 5 - 6 | 1154 | 87 | ||
XM_006238391.2 | 5 - 6 | 1154 | 87 | |||
XM_006238395.2 | XP_006238457.1 | 4 - 5 | 545 | 87 | ||
XM_008763848.1 | XP_008762070.1 | 4 - 5 | 860 | 87 | ||
XM_008763849.1 | XP_008762071.1 | 4 - 5 | 860 | 87 | ||
XM_008763850.1 | XP_008762072.1 | 4 - 5 | 523 | 87 | ||
GenBank mRNA | AB098713.1 | - | 5 - 6 | 1078 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
