Assay Details
Gene Symbol: Caap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034154.1 | NP_001029326.1 | 3 - 4 | 412 | 94 | |
XM_006238400.2 | XP_006238462.1 | 3 - 4 | 626 | 92 | ||
XM_008763894.1 | XP_008762116.1 | 3 - 4 | 410 | 92 | ||
GenBank mRNA | BC105829.1 | - | 3 - 4 | 412 | 7382013 | 94 |
Gene Symbol: LOC103692417 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC088436.1 | - | 442 | 92 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC105829.1, NM_001034154.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
