Assay Details
Gene Symbol: Angptl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025065.1 | NP_001020236.1 | 1 - 2 | 542 | 73 | |
XM_006238440.2 | XP_006238502.1 | 1 - 2 | 729 | 73 | ||
GenBank mRNA | BC088192.1 | - | 1 - 2 | 542 | 73 | |
FQ210243.1 | - | 1 - 2 | 552 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
