Assay Details
Gene Symbol: Pdxk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031769.1 | NP_113957.1 | 3 - 4 | 246 | 96 | |
GenBank mRNA | AF020346.1 | - | 3 - 4 | 246 | 96 | |
FQ213832.1 | - | 3 - 4 | 350 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
