Assay Details
Gene Symbol: Man1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033656.1 | NP_001028828.1 | 5 - 6 | 1422 | 96 | |
XM_006256497.1 | XP_006256559.1 | 5 - 6 | 1294 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
