Assay Details
Gene Symbol: Man1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033656.1 | NP_001028828.1 | 1 - 2 | 274 | 85 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001033656.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
