Assay Details
Gene Symbol: Lims1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145456.1 | NP_001138928.1 | 3 - 4 | 392 | 126 | |
XM_006256385.2 | XP_006256447.1 | 3 - 4 | 2294 | 126 | ||
XM_006256386.2 | XP_006256448.1 | 3 - 4 | 2294 | 126 | ||
XM_006256387.2 | XP_006256449.1 | 3 - 4 | 2291 | 126 | ||
XM_006256388.2 | XP_006256450.1 | 3 - 4 | 2158 | 126 | ||
XM_006256389.2 | XP_006256451.1 | 3 - 4 | 385 | 126 | ||
XM_006256390.2 | XP_006256452.1 | 3 - 4 | 1965 | 126 | ||
XM_006256391.2 | XP_006256453.1 | 3 - 4 | 1964 | 126 | ||
GenBank mRNA | FJ640984.1 | - | 3 - 4 | 392 | 126 | |
FJ640985.1 | - | 3 - 4 | 471 | 126 | ||
FQ222665.1 | - | 3 - 4 | 480 | 126 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
