Assay Details
Gene Symbol: Lims1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145456.1 | NP_001138928.1 | 4 - 5 | 515 | 94 | |
XM_006256385.2 | XP_006256447.1 | 4 - 5 | 2417 | 94 | ||
XM_006256386.2 | XP_006256448.1 | 4 - 5 | 2417 | 94 | ||
XM_006256387.2 | XP_006256449.1 | 4 - 5 | 2414 | 94 | ||
XM_006256388.2 | XP_006256450.1 | 4 - 5 | 2281 | 94 | ||
XM_006256389.2 | XP_006256451.1 | 4 - 5 | 508 | 94 | ||
XM_006256390.2 | XP_006256452.1 | 4 - 5 | 2088 | 94 | ||
XM_006256391.2 | XP_006256453.1 | 4 - 5 | 2087 | 94 | ||
GenBank mRNA | FJ640984.1 | - | 4 - 5 | 515 | 94 | |
FJ640985.1 | - | 4 - 5 | 594 | 94 | ||
FQ222665.1 | - | 4 - 5 | 603 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
