Assay Details
Gene Symbol: Lims1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145456.1 | NP_001138928.1 | 9 - 10 | 1037 | 83 | |
XM_006256387.2 | XP_006256449.1 | 9 - 10 | 2936 | 83 | ||
XM_006256391.2 | XP_006256453.1 | 9 - 10 | 2609 | 83 | ||
GenBank mRNA | FJ640984.1 | - | 9 - 10 | 1037 | 83 | |
FJ640985.1 | - | 9 - 10 | 1116 | 83 | ||
FQ222665.1 | - | 9 - 10 | 1125 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
epithelial to mesenchymal transition
cell-matrix adhesion
establishment or maintenance of cell polarity
positive regulation of gene expression
positive regulation of cell-substrate adhesion
chordate embryonic development
negative regulation of apoptotic process
positive regulation of GTPase activity
establishment of protein localization
cell-cell junction organization
negative regulation of transcription, DNA-templated
regulation of epithelial cell proliferation
negative regulation of epithelial cell proliferation
protein heterooligomerization
positive regulation of focal adhesion assembly
cellular response to transforming growth factor beta stimulus
cell-cell adhesion
positive regulation of substrate adhesion-dependent cell spreading
negative regulation of neural precursor cell proliferation
negative regulation of hepatocyte proliferation
positive regulation of integrin-mediated signaling pathway
zinc ion binding
protein kinase binding
protein domain specific binding
epithelial to mesenchymal transition
cell-matrix adhesion
establishment or maintenance of cell polarity
positive regulation of gene expression
positive regulation of cell-substrate adhesion
chordate embryonic development
negative regulation of apoptotic process
positive regulation of GTPase activity
establishment of protein localization
cell-cell junction organization
negative regulation of transcription, DNA-templated
regulation of epithelial cell proliferation
negative regulation of epithelial cell proliferation
protein heterooligomerization
positive regulation of focal adhesion assembly
cellular response to transforming growth factor beta stimulus
cell-cell adhesion
positive regulation of substrate adhesion-dependent cell spreading
negative regulation of neural precursor cell proliferation
negative regulation of hepatocyte proliferation
positive regulation of integrin-mediated signaling pathway
zinc ion binding
protein kinase binding
protein domain specific binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P457246 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731727 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450406 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496145 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376680 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 216830 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308654 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873145 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251357 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 180515 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673623 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575155 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271025 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722165 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335678 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399783 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 941042 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387873 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727058 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461480 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533755 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566363 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802401 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693418 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 927044 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641787 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720230 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 567428 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 695308 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 347398 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912838 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820147 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 105425 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283962 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341206 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818943 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890712 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 48688 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146346 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 175749 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 172385 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 73448 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77671 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114186 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 110464 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P457246 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P457246 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P457246 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P457246 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10833526 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10833534 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10833524 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10833529 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10833533 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10833531 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10833530 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10833527 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10833532 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10833528 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10833523 | - | - |
Rat Genome 230 2.0 Array | 1385430_PM_at | - | - |
Rat Genome 230 2.0 Array | 1385430_at | - | - |
Affymetrix HT Rat Focus Array | 1385430_at | - | - |
Affymetrix HT Rat Focus Array | 1385430_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR2000005542.rn.1 | TC2000000629.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR2000005541.rn.1 | TC2000000629.rn.1 | RTA TC |
Rat Expression Set 230 | 1385430_at | - | - |
Rat Expression Set 230 | 1385430_PM_at | - | - |
Rat Genome U34 Set | rc_AA799637_at | - | - |
Rat Genome U34 Set | rc_AA799637_g_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22153839 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1650346 | - | - |
