Assay Details
Gene Symbol: Fam184a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006256500.2 | XP_006256562.1 | 7 - 8 | 1668 | 105 | |
XM_008773011.1 | XP_008771233.1 | 7 - 8 | 1668 | 105 | ||
XM_008773012.1 | XP_008771234.1 | 7 - 8 | 1668 | 105 | ||
XM_008773013.1 | XP_008771235.1 | 7 - 8 | 1668 | 105 | ||
XM_008773014.1 | XP_008771236.1 | 7 - 8 | 1668 | 105 | ||
XM_008773015.1 | XP_008771237.1 | 8 - 9 | 1811 | 105 | ||
XM_008773016.1 | XP_008771238.1 | 8 - 9 | 1811 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
