Assay Details
Gene Symbol: Fam184a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006256500.2 | XP_006256562.1 | 8 - 9 | 1810 | 71 | |
XM_008773014.1 | XP_008771236.1 | 8 - 9 | 1810 | 71 | ||
XM_008773016.1 | XP_008771238.1 | 9 - 10 | 1953 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
