Assay Details
Gene Symbol: Fam184a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006256500.2 | XP_006256562.1 | 9 - 10 | 2025 | 61 | |
XM_008773011.1 | XP_008771233.1 | 10 - 11 | 2067 | 61 | ||
XM_008773012.1 | XP_008771234.1 | 10 - 11 | 2067 | 61 | ||
XM_008773013.1 | XP_008771235.1 | 10 - 11 | 2067 | 61 | ||
XM_008773014.1 | XP_008771236.1 | 9 - 10 | 2025 | 61 | ||
XM_008773015.1 | XP_008771237.1 | 11 - 12 | 2210 | 61 | ||
XM_008773016.1 | XP_008771238.1 | 10 - 11 | 2168 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
