Assay Details
Gene Symbol: Fam184a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008773011.1 | XP_008771233.1 | 13 - 14 | 2533 | 79 | |
XM_008773012.1 | XP_008771234.1 | 13 - 14 | 2533 | 79 | ||
XM_008773015.1 | XP_008771237.1 | 14 - 15 | 2676 | 79 | ||
XM_008773016.1 | XP_008771238.1 | 13 - 14 | 2634 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
