Assay Details
Gene Symbol: Pkib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076553.2 | NP_001070021.1 | 2 - 3 | 200 | 153 | |
NM_012627.3 | NP_036759.2 | 2 - 3 | 200 | 153 | ||
XM_006256512.2 | XP_006256574.1 | 3 - 4 | 322 | 153 | ||
XM_008772976.1 | XP_008771198.1 | 3 - 4 | 459 | 153 | ||
GenBank mRNA | AF413572.1 | - | 2 - 3 | 183 | 152 | |
BG666532.1 | - | 2 - 3 | 229 | 153 | ||
FQ231722.1 | - | 2 - 3 | 196 | 153 | ||
M64092.1 | - | 2 - 3 | 157 | 153 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF413572.1, M64092.1, NM_001076553.2, NM_012627.3, XM_006256512.2, XM_008772976.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
