Assay Details
Gene Symbol: Pkib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076553.2 | NP_001070021.1 | 4 - 5 | 371 | 158 | |
XM_006256511.2 | XP_006256573.1 | 4 - 5 | 990 | 158 | ||
XM_006256512.2 | XP_006256574.1 | 5 - 6 | 493 | 158 | ||
XM_006256513.2 | XP_006256575.1 | 4 - 5 | 348 | 158 | ||
XM_006256515.2 | XP_006256577.1 | 4 - 5 | 913 | 158 | ||
XM_008772976.1 | XP_008771198.1 | 5 - 6 | 630 | 158 | ||
XM_008772977.1 | XP_008771199.1 | 5 - 6 | 1343 | 158 | ||
XM_008772978.1 | XP_008771200.1 | 4 - 5 | 1080 | 158 | ||
GenBank mRNA | BG668634.1 | - | 438 | 158 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001076553.2, XM_006256511.2, XM_006256512.2, XM_006256513.2, XM_006256515.2, XM_008772976.1, XM_008772977.1, XM_008772978.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
