Assay Details
Gene Symbol: Gopc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107631.1 | NP_001101101.1 | 2 - 3 | 617 | 103 | |
GenBank mRNA | BC168160.1 | - | 2 - 3 | 646 | 103 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC168160.1, NM_001107631.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
