Assay Details
Gene Symbol: Gopc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107631.1 | NP_001101101.1 | 4 - 5 | 815 | 104 | |
GenBank mRNA | BC168160.1 | - | 4 - 5 | 844 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
