Assay Details
Gene Symbol: Gopc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107631.1 | NP_001101101.1 | 8 - 9 | 1431 | 78 | |
GenBank mRNA | BC168160.1 | - | 8 - 9 | 1460 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P422063 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89346 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507087 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151506 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144490 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910915 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297701 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 791505 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692615 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345645 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734335 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837506 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546909 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 316535 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 227326 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 114869 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965189 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541083 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 973458 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566174 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854913 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275693 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544610 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814086 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 771073 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21203 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491744 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268951 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250532 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 430165 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318693 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 798954 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 165906 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62587 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 122281 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 92330 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149994 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32477 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P422063 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P422063 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P422063 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P422063 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P977419 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P929979 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 53280 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10833340 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10833339 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10833341 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10833342 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10833338 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10833343 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10833345 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10833344 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10833337 | - | - |
Rat Genome 230 2.0 Array | 1391928_at | - | - |
Rat Genome 230 2.0 Array | 1391183_at | - | - |
Rat Genome 230 2.0 Array | 1391183_PM_at | - | - |
Rat Genome 230 2.0 Array | 1391928_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR2000013070.rn.1 | TC2000001585.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR2000013069.rn.1 | TC2000001585.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR2000013066.rn.1 | TC2000001585.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR2000013067.rn.1 | TC2000001585.rn.1 | RTA TC |
Rat Expression Set 230 | 1391928_at | - | - |
Rat Expression Set 230 | 1391183_PM_at | - | - |
Rat Expression Set 230 | 1391928_PM_at | - | - |
Rat Expression Set 230 | 1391183_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10833337 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22075806 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1365955 | - | - |
