Assay Details
Gene Symbol: Dnajb12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013907.1 | NP_001013929.1 | 4 - 5 | 763 | 110 | |
XM_006256411.2 | XP_006256473.1 | 4 - 5 | 776 | 110 | ||
XM_006256412.2 | XP_006256474.1 | 4 - 5 | 776 | 110 | ||
GenBank mRNA | BC090076.1 | - | 4 - 5 | 763 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
