Assay Details
Gene Symbol: Aifm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001139483.1 | NP_001132955.1 | 3 - 4 | 438 | 67 | |
XM_006256452.2 | XP_006256514.1 | 4 - 5 | 552 | 67 | ||
XM_006256453.2 | XP_006256515.1 | 4 - 5 | 552 | 67 | ||
XM_006256454.2 | XP_006256516.1 | 4 - 5 | 495 | 67 | ||
XM_006256456.2 | XP_006256518.1 | 4 - 5 | 522 | 67 | ||
XM_006256459.2 | XP_006256521.1 | 4 - 5 | 552 | 67 | ||
XM_008772888.1 | XP_008771110.1 | 4 - 5 | 552 | 67 | ||
XM_008772889.1 | XP_008771111.1 | 4 - 5 | 510 | 67 | ||
XM_008772890.1 | XP_008771112.1 | 4 - 5 | 497 | 67 | ||
GenBank mRNA | AW521424.1 | - | 3 - 4 | 341 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
