Assay Details
Gene Symbol: Actr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009268.1 | NP_001009268.1 | 4 - 5 | 511 | 71 | |
GenBank mRNA | BC088442.1 | - | 4 - 5 | 511 | 7126325 | 71 |
FQ212547.1 | - | 4 - 5 | 510 | 71 | ||
FQ212814.1 | - | 5 - 6 | 475 | 71 | ||
FQ219071.1 | - | 4 - 5 | 460 | 71 | ||
FQ219838.1 | - | 4 - 5 | 477 | 71 | ||
FQ225424.1 | - | 4 - 5 | 482 | 71 | ||
FQ226083.1 | - | 4 - 5 | 457 | 71 | ||
FQ226422.1 | - | 4 - 5 | 460 | 71 | ||
FQ227488.1 | - | 4 - 5 | 509 | 71 | ||
FQ227830.1 | - | 4 - 5 | 460 | 71 | ||
FQ230874.1 | - | 4 - 5 | 495 | 71 | ||
FQ231828.1 | - | 4 - 5 | 462 | 71 | ||
FQ231874.1 | - | 4 - 5 | 495 | 71 | ||
FQ234230.1 | - | 4 - 5 | 462 | 71 | ||
FQ235071.1 | - | 4 - 5 | 482 | 71 | ||
FQ235201.1 | - | 4 - 5 | 477 | 71 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
