Assay Details
Gene Symbol: Actr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009268.1 | NP_001009268.1 | 6 - 7 | 806 | 94 | |
GenBank mRNA | BC088442.1 | - | 6 - 7 | 806 | 94 | |
FQ212547.1 | - | 6 - 7 | 805 | 94 | ||
FQ212814.1 | - | 7 - 8 | 770 | 94 | ||
FQ219838.1 | - | 6 - 7 | 772 | 94 | ||
FQ225424.1 | - | 6 - 7 | 777 | 94 | ||
FQ226083.1 | - | 6 - 7 | 752 | 94 | ||
FQ226422.1 | - | 6 - 7 | 755 | 94 | ||
FQ227142.1 | - | 6 - 7 | 755 | 94 | ||
FQ227488.1 | - | 6 - 7 | 805 | 94 | ||
FQ227830.1 | - | 6 - 7 | 755 | 94 | ||
FQ230292.1 | - | 2 - 3 | 298 | 94 | ||
FQ231828.1 | - | 6 - 7 | 757 | 94 | ||
FQ231874.1 | - | 6 - 7 | 790 | 94 | ||
FQ234230.1 | - | 6 - 7 | 757 | 94 | ||
FQ235071.1 | - | 6 - 7 | 777 | 94 | ||
FQ235201.1 | - | 6 - 7 | 772 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
