Assay Details
Gene Symbol: Ugp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024743.1 | NP_001019914.1 | 2 - 3 | 396 | 95 | |
XM_006251538.1 | XP_006251600.1 | 2 - 3 | 348 | 95 | ||
GenBank mRNA | BC097369.1 | - | 2 - 3 | 396 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
