Assay Details
Gene Symbol: Ugp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024743.1 | NP_001019914.1 | 6 - 7 | 1116 | 87 | |
XM_006251538.1 | XP_006251600.1 | 6 - 7 | 1068 | 87 | ||
GenBank mRNA | BC084711.1 | - | 1 - 2 | 223 | 87 | |
BC097369.1 | - | 6 - 7 | 1116 | 87 | ||
FQ214781.1 | - | 1 - 2 | 299 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
