Assay Details
Gene Symbol: Fancl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191684.1 | NP_001178613.1 | 3 - 4 | 222 | 158 | |
XM_006251597.2 | XP_006251659.1 | 3 - 4 | 990 | 158 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
