Assay Details
Gene Symbol: Peli1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100565.1 | NP_001094035.1 | 4 - 5 | 585 | 107 | |
GenBank mRNA | BC166745.1 | - | 4 - 5 | 418 | 7939059 | 107 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
