Assay Details
Gene Symbol: Peli1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100565.1 | NP_001094035.1 | 6 - 7 | 972 | 62 | |
GenBank mRNA | BC166745.1 | - | 6 - 7 | 805 | 7939059 | 62 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein polyubiquitination
positive regulation of cytokine production
Toll signaling pathway
biological_process
positive regulation of B cell proliferation
positive regulation of protein ubiquitination
negative regulation of NF-kappaB transcription factor activity
response to lipopolysaccharide
positive regulation of toll-like receptor 3 signaling pathway
positive regulation of toll-like receptor 4 signaling pathway
negative regulation of T cell proliferation
positive regulation of I-kappaB kinase/NF-kappaB signaling
response to dsRNA
negative regulation of T cell activation
positive regulation of B cell activation
protein K48-linked ubiquitination
molecular_function
ubiquitin-protein transferase activity
ubiquitin protein ligase activity
protein polyubiquitination
positive regulation of cytokine production
Toll signaling pathway
biological_process
positive regulation of B cell proliferation
positive regulation of protein ubiquitination
negative regulation of NF-kappaB transcription factor activity
response to lipopolysaccharide
positive regulation of toll-like receptor 3 signaling pathway
positive regulation of toll-like receptor 4 signaling pathway
negative regulation of T cell proliferation
positive regulation of I-kappaB kinase/NF-kappaB signaling
response to dsRNA
negative regulation of T cell activation
positive regulation of B cell activation
protein K48-linked ubiquitination
molecular_function
ubiquitin-protein transferase activity
ubiquitin protein ligase activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P155826 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206199 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504138 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230985 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 1126 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353108 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445443 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 800044 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 466347 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 530148 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 424067 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685980 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 926559 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936251 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 700496 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10841 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 136108 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57820 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654995 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777149 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 743031 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90893 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603004 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349283 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 4479 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 923978 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464023 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692864 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 487882 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673965 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892526 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275781 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855102 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 774630 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816292 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680573 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591881 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716850 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963950 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318597 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343990 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 153746 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 42891 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 45247 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39506 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 76027 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121653 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19187 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P155826 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P155826 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P155826 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P243426 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P341456 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10774381 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10774380 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10774382 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10774377 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10774379 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10774378 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10774375 | - | - |
Rat Genome 230 2.0 Array | 1399081_at | - | - |
Rat Genome 230 2.0 Array | 1390814_at | - | - |
Rat Genome 230 2.0 Array | 1399081_PM_at | - | - |
Rat Genome 230 2.0 Array | 1390814_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1390814_at | - | - |
Affymetrix HT Rat Focus Array | 1390814_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1400007934.rn.1 | TC1400001101.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1400007935.rn.1 | TC1400001101.rn.1 | RTA TC |
Rat Expression Set 230 | 1399081_PM_at | - | - |
Rat Expression Set 230 | 1399081_at | - | - |
Rat Expression Set 230 | 1390814_PM_at | - | - |
Rat Expression Set 230 | 1390814_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10774375 | - | - |
Rat Genome U34 Set | rc_AA899973_at | - | - |
Rat Genome U34 Set | rc_AA848897_at | - | - |
Rat Genome U34 Set | rc_AI112111_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1371904 | - | - |
