Assay Details
Gene Symbol: Efemp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012039.1 | NP_001012039.1 | 9 - 10 | 1101 | 62 | |
XM_008770456.1 | XP_008768678.1 | 9 - 10 | 1201 | 62 | ||
XM_008770457.1 | XP_008768679.1 | 8 - 9 | 1121 | 62 | ||
GenBank mRNA | BC079455.1 | - | 9 - 10 | 1101 | 62 | |
D89730.1 | - | 7 - 8 | 997 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
