Assay Details
Gene Symbol: Efemp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012039.1 | NP_001012039.1 | 10 - 11 | 1231 | 145 | |
XM_008770456.1 | XP_008768678.1 | 10 - 11 | 1331 | 145 | ||
XM_008770457.1 | XP_008768679.1 | 9 - 10 | 1251 | 145 | ||
GenBank mRNA | BC079455.1 | - | 10 - 11 | 1231 | 7121509 | 145 |
D89730.1 | - | 8 - 9 | 1127 | 145 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
