Assay Details
Gene Symbol: Ppp3r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017309.2 | NP_059005.1 | 4 - 5 | 491 | 65 | |
XM_006251517.2 | XP_006251579.2 | 4 - 5 | 534 | 65 | ||
XM_008770426.1 | XP_008768648.1 | 3 - 4 | 1204 | 65 | ||
GenBank mRNA | BC088855.1 | - | 4 - 5 | 491 | 65 | |
D14425.1 | - | 4 - 5 | 459 | 65 | ||
D14568.1 | - | 4 - 5 | 347 | 65 | ||
FQ212066.1 | - | 4 - 5 | 658 | 65 | ||
L03554.1 | - | 3 - 4 | 293 | 65 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
