Assay Details
Gene Symbol: Dusp22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108412.1 | NP_001101882.1 | 2 - 3 | 171 | 95 | |
XM_006253888.2 | XP_006253950.1 | 2 - 3 | 119 | 95 | ||
XM_006253889.2 | XP_006253951.1 | 2 - 3 | 392 | 95 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
