Assay Details
Gene Symbol: Nqo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004214.1 | NP_001004214.1 | 2 - 3 | 113 | 102 | |
XM_006253867.2 | XP_006253929.1 | 2 - 3 | 364 | 102 | ||
GenBank mRNA | BC079157.1 | - | 2 - 3 | 113 | 102 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC079157.1, NM_001004214.1, XM_006253867.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
