Assay Details
Gene Symbol: Nqo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004214.1 | NP_001004214.1 | 3 - 4 | 275 | 139 | |
XM_006253867.2 | XP_006253929.1 | 3 - 4 | 526 | 139 | ||
XM_006253868.2 | XP_006253930.1 | 2 - 3 | 284 | 139 | ||
GenBank mRNA | BC079157.1 | - | 3 - 4 | 275 | 7128990 | 139 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
