Assay Details
Gene Symbol: Ptpdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106104.1 | NP_001099574.1 | 6 - 7 | 863 | 116 | |
XM_006253700.2 | XP_006253762.1 | 5 - 6 | 1686 | 116 | ||
XM_006253701.2 | XP_006253763.1 | 6 - 7 | 1383 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
