Assay Details
Gene Symbol: Ptpdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106104.1 | NP_001099574.1 | 9 - 10 | 2404 | 117 | |
XM_006253700.2 | XP_006253762.1 | 8 - 9 | 3227 | 117 | ||
XM_006253701.2 | XP_006253763.1 | 9 - 10 | 2924 | 117 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P538970 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553290 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499037 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 384609 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 131281 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 874986 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 81005 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 531796 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 564189 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304737 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890633 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 442011 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819159 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83611 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720702 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60019 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828334 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901170 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 596550 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240351 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853658 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197197 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472511 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362932 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 292129 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387137 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 282225 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 165971 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 570594 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342202 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 747345 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160141 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 762676 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708451 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 967244 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 287519 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 105873 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70043 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 60083 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 83145 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P538970 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P538970 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P538970 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P538970 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10794431 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10794438 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10794440 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10794439 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10794441 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10794432 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10794435 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10794434 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10794436 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10794437 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10794430 | - | - |
Affymetrix Rat Clariom S Assay | PSR1700002344.rn.1 | TC1700000290.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1700002343.rn.1 | TC1700000290.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1700002345.rn.1 | TC1700000290.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1700002348.rn.1 | TC1700000290.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10794430 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22220911 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1367657 | - | - |
