Assay Details
Gene Symbol: Ippk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008556.1 | NP_001008556.1 | 6 - 7 | 501 | 110 | |
XM_008771534.1 | XP_008769756.1 | 5 - 6 | 1216 | 110 | ||
XM_008771535.1 | XP_008769757.1 | 6 - 7 | 600 | 110 | ||
GenBank mRNA | AY823319.1 | - | 6 - 7 | 501 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
