Assay Details
Gene Symbol: Ssr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008891.2 | NP_001008891.2 | 4 - 5 | 617 | 87 | |
XM_006253835.2 | XP_006253897.1 | 4 - 5 | 654 | 87 | ||
GenBank mRNA | AY321345.1 | - | 591 | 87 | ||
BC098008.1 | - | 4 - 5 | 617 | 87 | ||
FQ220080.1 | - | 4 - 5 | 585 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
