Assay Details
Gene Symbol: Ssr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008891.2 | NP_001008891.2 | 5 - 6 | 689 | 78 | |
XM_006253835.2 | XP_006253897.1 | 5 - 6 | 726 | 78 | ||
GenBank mRNA | AY321345.1 | - | 663 | 78 | ||
BC098008.1 | - | 5 - 6 | 689 | 78 | ||
FQ220080.1 | - | 5 - 6 | 657 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
