Assay Details
Gene Symbol: Wrnip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172332.2 | NP_758835.1 | 2 - 3 | 1173 | 156 | |
GenBank mRNA | AY136827.1 | - | 2 - 3 | 1077 | 156 | |
BC098652.1 | - | 2 - 3 | 1173 | 156 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
