Assay Details
Gene Symbol: Auh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108407.1 | NP_001101877.1 | 3 - 4 | 274 | 67 | |
XM_006253674.2 | XP_006253736.1 | 3 - 4 | 372 | 67 | ||
XM_006253675.2 | XP_006253737.1 | 3 - 4 | 375 | 67 | ||
XM_008771499.1 | XP_008769721.1 | 3 - 4 | 1328 | 67 | ||
GenBank mRNA | FQ218844.1 | - | 3 - 4 | 366 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
