Assay Details
Gene Symbol: Auh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108407.1 | NP_001101877.1 | 9 - 10 | 792 | 126 | |
XM_006253674.2 | XP_006253736.1 | 9 - 10 | 890 | 126 | ||
XM_006253675.2 | XP_006253737.1 | 8 - 9 | 800 | 126 | ||
XM_008771499.1 | XP_008769721.1 | 9 - 10 | 1846 | 126 | ||
GenBank mRNA | FQ219791.1 | - | 4 - 5 | 476 | 126 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P16992 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294795 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 787501 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694157 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565309 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 96464 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 803744 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674209 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 53260 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354916 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 28474 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858644 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 163060 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676916 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 705714 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 767211 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90053 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 319132 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630209 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865499 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 632338 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326087 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865680 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 274642 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562949 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 33038 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 443012 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 513655 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 808728 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649407 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256961 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908678 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 651222 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 841422 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 161519 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258894 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921038 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744131 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945666 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 598451 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 725605 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324673 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854537 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519268 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 686310 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903953 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 479717 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488592 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681127 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 723124 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318579 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 293381 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785528 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603162 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 642962 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 971713 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294504 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 550736 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868707 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322015 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 547406 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407337 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 934839 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404749 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 455652 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898560 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 388879 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907807 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 20773 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 735286 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 209462 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79812 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 56112 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 116994 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 66666 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 107069 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 111649 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 168314 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 173314 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 1844 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 23516 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 138884 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P16992 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P16992 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P16992 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P16992 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 6626 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 45284 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 82126 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 45751 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10794237 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10794236 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10794235 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10794233 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10794238 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10794239 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10794231 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10794232 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10794234 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10794229 | - | - |
Rat Genome 230 2.0 Array | 1372149_PM_at | - | - |
Rat Genome 230 2.0 Array | 1372149_at | - | - |
Affymetrix HT Rat Focus Array | 1372149_at | - | - |
Affymetrix HT Rat Focus Array | 1372149_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1700001775.rn.1 | TC1700000215.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1700001776.rn.1 | TC1700000215.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1700001772.rn.1 | TC1700000215.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1700001774.rn.1 | TC1700000215.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1700001777.rn.1 | TC1700000215.rn.1 | RTA TC |
Rat Expression Set 230 | 1372149_PM_at | - | - |
Rat Expression Set 230 | 1372149_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10794229 | - | - |
Rat Genome U34 Set | rc_AA859688_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21123413 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1366845 | - | - |
