Assay Details
Gene Symbol: Auh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108407.1 | NP_001101877.1 | 2 - 3 | 190 | 111 | |
XM_006253674.2 | XP_006253736.1 | 2 - 3 | 288 | 111 | ||
XM_006253675.2 | XP_006253737.1 | 2 - 3 | 291 | 111 | ||
XM_008771499.1 | XP_008769721.1 | 2 - 3 | 1244 | 111 | ||
GenBank mRNA | FQ218844.1 | - | 2 - 3 | 282 | 111 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008771499.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
