Assay Details
Gene Symbol: Slc22a23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022624.1 | NP_072146.1 | 6 - 7 | 2055 | 84 | |
XM_006253845.2 | XP_006253907.1 | 6 - 7 | 1335 | 84 | ||
GenBank mRNA | AF184921.1 | - | 3 - 4 | 323 | 84 | |
BC158843.1 | - | 6 - 7 | 954 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
