Assay Details
Gene Symbol: Slc22a23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022624.1 | NP_072146.1 | 10 - 11 | 2467 | 120 | |
XM_006253845.2 | XP_006253907.1 | 9 - 10 | 1723 | 120 | ||
GenBank mRNA | AF184921.1 | - | 6 - 7 | 711 | 120 | |
BC158843.1 | - | 9 - 10 | 1342 | 120 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P318546 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P732101 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 443909 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657779 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 689475 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 136359 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698562 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683992 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270956 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 53475 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 509813 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56330 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 817695 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393931 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380416 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 862275 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 510545 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589040 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465090 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 931157 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 162392 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297508 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 874280 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 371478 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 794764 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698617 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736190 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 666224 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273549 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289239 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819064 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378285 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 530730 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107117 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 973409 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488635 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236861 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929791 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144115 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518098 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669358 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393329 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 686559 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 40230 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 927072 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568394 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 788091 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259117 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575512 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189234 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 53783 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919269 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 474196 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 269880 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 425550 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 371140 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75148 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372869 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110432 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138287 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726047 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853162 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702048 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 592617 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621533 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594435 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 156298 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795054 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285377 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12630 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793549 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 239709 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 156118 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 8105 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 739658 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408503 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413764 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249471 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843620 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913228 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291447 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30491 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 125161 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437399 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 94097 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 46642 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871451 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 455466 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 428153 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 646571 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 510076 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70687 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389275 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 956642 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 651695 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499654 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422029 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861324 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284006 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 900249 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 443879 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617319 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354876 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 32576 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709887 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109515 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 523537 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851174 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473766 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851098 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884465 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214302 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491770 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595596 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 841223 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 41578 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933050 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63250 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 420936 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6804 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 760702 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226709 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893900 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 136672 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19126 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 108403 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28620 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47315 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 82334 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 138270 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 64587 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 148060 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 116089 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 131076 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 97353 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 131058 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 166519 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 136847 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 49656 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 135181 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 148082 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P318546 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P732101 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P732101 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P318546 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P318546 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P732101 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P318546 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 702124175 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 87542 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 47480 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 65896 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 10134 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 93831 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 78755 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 702124175 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10794819 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10794818 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10794820 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10794817 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10794814 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10794813 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10794816 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10794815 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10794822 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10794823 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10794821 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10794812 | - | - |
Rat Genome 230 2.0 Array | 1376860_at | - | - |
Rat Genome 230 2.0 Array | 1371069_PM_at | - | - |
Rat Genome 230 2.0 Array | 1371069_at | - | - |
Rat Genome 230 2.0 Array | 1376860_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1371069_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1371069_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1700003482.rn.1 | TC1700000487.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1700003476.rn.1 | TC1700000487.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1700003481.rn.1 | TC1700000487.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1700003479.rn.1 | TC1700000487.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1700003480.rn.1 | TC1700000487.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1700003478.rn.1 | TC1700000487.rn.1 | RTA TC |
Rat Expression Set 230 | 1371069_PM_at | - | - |
Rat Expression Set 230 | 1376860_PM_at | - | - |
Rat Expression Set 230 | 1376860_at | - | - |
Rat Expression Set 230 | 1371069_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10794812 | - | - |
Rat Genome U34 Set | rc_AA997237_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21202480 | - | - |
ABI Rat Genome Survey Microarray | 22335818 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1369142 | - | - |
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1530318 | - | - |
